Assay Details
| Gene Symbol: ZNF500 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001098209.4 | XP_001098209.2 | 716 | 95 | ||
| XM_015125553.2 | XP_014981039.2 | 843 | 95 | |||
| XM_028840683.1 | XP_028696516.1 | 576 | 95 | |||
| GenBank mRNA | JV647875.1 | - | 130 | 95 | ||
More Information
Set Membership: |
|

ZNF500
Genomic Map