Assay Details
| Gene Symbol: TLNRD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001194462.2 | NP_001181391.2 | 901 | 105 | ||
| GenBank mRNA | JU322205.1 | - | 499 | 105 | ||
| JU322206.1 | - | 499 | 105 | |||
| JU473140.1 | - | 444 | 105 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001194462.2 transcripts.
Set Membership: |
|

TLNRD1
Genomic Map