Assay Details
| Gene Symbol: ASB7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001266289.2 | NP_001253218.1 | 757 | 77 | ||
| GenBank mRNA | JU333018.1 | - | 920 | 77 | ||
| JU474371.1 | - | 757 | 77 | |||
| JV642740.1 | - | 967 | 77 | |||
More Information
Set Membership: |
|

ASB7
Genomic Map