Assay Details
| Gene Symbol: WNT5B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015150770.2 | XP_015006256.1 | 1596 | 121 | ||
| GenBank mRNA | JV662474.1 | - | 132 | 121 | ||
| JV724240.1 | - | 121 | ||||
More Information
Set Membership: |
|

WNT5B
Genomic Map