Assay Details
| Gene Symbol: GFOD2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001260710.1 | NP_001247639.1 | 489 | 76 | ||
| XM_028840426.1 | XP_028696259.1 | 816 | 76 | |||
| XM_028840427.1 | XP_028696260.1 | 935 | 76 | |||
| XM_028840428.1 | XP_028696261.1 | 847 | 76 | |||
| GenBank mRNA | JU336542.1 | - | 508 | 76 | ||
| JU473181.1 | - | 472 | 76 | |||
| JV048339.1 | - | 489 | 76 | |||
| JV629981.1 | - | 524 | 76 | |||
More Information
Set Membership: |
|

GFOD2
Genomic Map