Assay Details
Gene Symbol: CCDC8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001111790.4 | XP_001111790.3 | 728 | 145 | ||
GenBank mRNA | JV073002.1 | - | 702 | 145 | ||
JV663067.1 | - | 688 | 145 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_001111790.4 transcripts.
Set Membership: |
![]() ![]() ![]() |
