Assay Details
| Gene Symbol: TMEM191C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001193786.2 | NP_001180715.1 | 552 | 134 | ||
| XM_015149747.2 | XP_015005233.1 | 722 | 134 | |||
| XM_015149748.2 | XP_015005234.1 | 674 | 134 | |||
| XM_015149749.2 | XP_015005235.1 | 716 | 134 | |||
More Information
Set Membership: |
|

TMEM191C
Genomic Map