Assay Details
| Gene Symbol: FCRLA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001118111.4 | XP_001118111.2 | 798 | 150 | ||
| XM_015113959.2 | XP_014969445.1 | 802 | 150 | |||
| XM_015113964.2 | XP_014969450.1 | 813 | 150 | |||
| GenBank mRNA | JU495078.1 | - | 553 | 150 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

FCRLA
Genomic Map