Assay Details
| Gene Symbol: SMDT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001194351.1 | NP_001181280.1 | 254 | 95 | ||
| GenBank mRNA | JU323412.1 | - | 245 | 95 | ||
| JU472913.1 | - | 264 | 95 | |||
| JV453032.1 | - | 264 | 95 | |||
| JV669732.1 | - | 260 | 95 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

SMDT1
Genomic Map