Assay Details
| Gene Symbol: RWDD2A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001084182.4 | XP_001084182.2 | 475 | 141 | ||
| GenBank mRNA | JU320179.1 | - | 378 | 141 | ||
| JV529002.1 | - | 423 | 141 | |||
| JV704470.1 | - | 399 | 141 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

RWDD2A
Genomic Map