Assay Details
| Gene Symbol: NANP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001266157.1 | NP_001253086.1 | 127 | 122 | ||
| GenBank mRNA | JU326715.1 | - | 88 | 122 | ||
| JU472019.1 | - | 124 | 122 | |||
| JV045271.1 | - | 135 | 122 | |||
| JV635804.1 | - | 127 | 122 | |||
More Information
Set Membership: |
|

NANP
Genomic Map