Assay Details
| Gene Symbol: LRRC57 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001194887.2 | NP_001181816.1 | 343 | 109 | ||
| XM_015141911.2 | XP_014997397.1 | 444 | 109 | |||
| GenBank mRNA | JU333810.1 | - | 328 | 109 | ||
| JU333811.1 | - | 328 | 109 | |||
| JU333812.1 | - | 328 | 109 | |||
| JU333813.1 | - | 328 | 109 | |||
| JU475035.1 | - | 332 | 109 | |||
More Information
Set Membership: |
|

LRRC57
Genomic Map