Assay Details
| Gene Symbol: C1QC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001266808.1 | NP_001253737.1 | 246 | 104 | ||
| XM_015130170.2 | XP_014985656.1 | 324 | 104 | |||
| GenBank mRNA | JU323143.1 | - | 267 | 104 | ||
| JU472221.1 | - | 246 | 104 | |||
| JV490269.1 | - | 286 | 104 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

C1QC
Genomic Map