Assay Details
| Gene Symbol: SPPL2C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001115879.4 | XP_001115879.2 | 811 | 156 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

SPPL2C| Gene Symbol: SPPL2C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001115879.4 | XP_001115879.2 | 811 | 156 | ||
Set Membership: |
|