Assay Details
| Gene Symbol: PRPS1L1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001105054.4 | XP_001105054.2 | 74 | 101 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_001105054.4 transcripts.
Set Membership: |
|

PRPS1L1
Genomic Map