Assay Details
| Gene Symbol: DHRS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001109672.4 | XP_001109672.2 | 705 | 160 | ||
| XM_028851433.1 | XP_028707266.1 | 705 | 160 | |||
| XM_028851434.1 | XP_028707267.1 | 312 | 160 | |||
| GenBank mRNA | JV639868.1 | - | 235 | 160 | ||
More Information
Set Membership: |
|

DHRS2
Genomic Map