Assay Details
| Gene Symbol: F9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001109683.1 | NP_001103153.1 | 864 | 81 | ||
| XM_028841666.1 | XP_028697499.1 | 813 | 81 | |||
| XM_028841667.1 | XP_028697500.1 | 744 | 81 | |||
| GenBank mRNA | EU128170.1 | - | 864 | 81 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

F9
Genomic Map