Assay Details
| Gene Symbol: STAR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001265769.1 | NP_001252698.1 | 519 | 86 | ||
| GenBank mRNA | JV044226.1 | - | 482 | 86 | ||
| JV635390.1 | - | 519 | 86 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

STAR
Genomic Map