Assay Details
| Gene Symbol: TCN2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001110286.4 | XP_001110286.2 | 580 | 95 | ||
| XM_015150160.2 | XP_015005646.2 | 577 | 95 | |||
| XM_028827842.1 | XP_028683675.1 | 757 | 95 | |||
| GenBank mRNA | JV452569.1 | - | 373 | 95 | ||
More Information
Set Membership: |
|

TCN2
Genomic Map