Assay Details
| Gene Symbol: CRYBB2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001122894.2 | NP_001116366.2 | 220 | 101 | ||
| GenBank mRNA | EG703254.1 | - | 226 | 101 | ||
| EG705486.1 | - | 194 | 101 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

CRYBB2
Genomic Map