Assay Details
| Gene Symbol: DCK | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001257554.1 | NP_001244483.1 | 151 | 96 | ||
| GenBank mRNA | JU323085.1 | - | 151 | 96 | ||
| JU472132.1 | - | 353 | 96 | |||
| JV044117.1 | - | 297 | 96 | |||
More Information
Set Membership: |
|

DCK
Genomic Map