Assay Details
| Gene Symbol: IFNAR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001092342.4 | XP_001092342.2 | 445 | 139 | ||
| XM_015132995.2 | XP_014988481.1 | 595 | 139 | |||
| XM_015132996.2 | XP_014988482.1 | 594 | 139 | |||
| XM_015132998.2 | XP_014988484.1 | 594 | 139 | |||
| XM_015132999.2 | XP_014988485.1 | 594 | 139 | |||
| GenBank mRNA | JU334340.1 | - | 426 | 139 | ||
| JU475438.1 | - | 292 | 139 | |||
| JU475439.1 | - | 292 | 139 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

IFNAR2
Genomic Map