Assay Details
| Gene Symbol: PRIM2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001266600.1 | NP_001253529.1 | 1286 | 95 | ||
| GenBank mRNA | JU322951.1 | - | 1308 | 95 | ||
| JU335349.1 | - | 1308 | 95 | |||
| JU476310.1 | - | 1286 | 95 | |||
| JU476311.1 | - | 1286 | 95 | |||
| JU476312.1 | - | 1286 | 95 | |||
| JV655544.1 | - | 757 | 95 | |||
More Information
Set Membership: |
|

PRIM2
Genomic Map