Assay Details
| Gene Symbol: PRSS58 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001085477.4 | XP_001085477.2 | 811 | 146 | ||
| XM_015135104.2 | XP_014990590.2 | 699 | 146 | |||
More Information
Set Membership: |
|

PRSS58
Genomic Map