Assay Details
| Gene Symbol: SMAD5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001266315.1 | NP_001253244.1 | 575 | 154 | ||
| XM_015140869.2 | XP_014996355.1 | 693 | 154 | |||
| XM_015140871.2 | XP_014996357.1 | 768 | 154 | |||
| XM_015140872.2 | XP_014996358.1 | 1263 | 154 | |||
| XM_028849266.1 | XP_028705099.1 | 667 | 154 | |||
| GenBank mRNA | JU334508.1 | - | 671 | 154 | ||
| JU334509.1 | - | 671 | 154 | |||
| JU334510.1 | - | 671 | 154 | |||
| JU334511.1 | - | 671 | 154 | |||
| JU334512.1 | - | 671 | 154 | |||
| JU334513.1 | - | 671 | 154 | |||
| JU334514.1 | - | 671 | 154 | |||
| JU472412.1 | - | 575 | 154 | |||
More Information
Set Membership: |
|

SMAD5
Genomic Map