Assay Details
| Gene Symbol: NFKBIB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001258157.1 | NP_001245086.1 | 672 | 127 | ||
| XM_015123803.2 | XP_014979289.1 | 539 | 127 | |||
| GenBank mRNA | JU329873.1 | - | 619 | 127 | ||
| JU470940.1 | - | 672 | 127 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

NFKBIB
Genomic Map