Assay Details
| Gene Symbol: STUM | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001266334.1 | NP_001253263.1 | 293 | 67 | ||
| XM_028831397.1 | XP_028687230.1 | 334 | 67 | |||
| GenBank mRNA | JV544104.1 | - | 307 | 67 | ||
| JV615932.1 | - | 313 | 67 | |||
| JV667393.1 | - | 293 | 67 | |||
More Information
Set Membership: |
|

STUM
Genomic Map