Assay Details
| Gene Symbol: RABL2B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001260670.2 | NP_001247599.1 | 369 | 135 | ||
| XM_015150634.2 | XP_015006120.2 | 732 | 135 | |||
| XM_015150635.2 | XP_015006121.2 | 842 | 135 | |||
| XM_015150638.2 | XP_015006124.2 | 486 | 135 | |||
| XM_028827398.1 | XP_028683231.1 | 990 | 135 | |||
| XM_028827399.1 | XP_028683232.1 | 986 | 135 | |||
| XM_028827400.1 | XP_028683233.1 | 704 | 135 | |||
| XM_028827401.1 | XP_028683234.1 | 595 | 135 | |||
| XM_028827402.1 | XP_028683235.1 | 880 | 135 | |||
| XM_028827403.1 | XP_028683236.1 | 726 | 135 | |||
| XM_028827404.1 | XP_028683237.1 | 267 | 135 | |||
| XM_028827405.1 | XP_028683238.1 | 880 | 135 | |||
| GenBank mRNA | JU323199.1 | - | 452 | 135 | ||
| JU471428.1 | - | 377 | 135 | |||
| JV046060.1 | - | 369 | 135 | |||
| JV567305.1 | - | 515 | 135 | |||
| JV704339.1 | - | 596 | 135 | |||
| Gene Symbol: LOC114677631 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_028847684.1 | XP_028703517.1 | 405 | 135 | ||
More Information
Set Membership: |
|

RABL2B
Genomic Map