Assay Details
| Gene Symbol: AGMO | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001105641.4 | XP_001105641.1 | 352 | 98 | ||
| XM_028845993.1 | XP_028701826.1 | 420 | 98 | |||
| XM_028845994.1 | XP_028701827.1 | 420 | 98 | |||
| XM_028845995.1 | XP_028701828.1 | 420 | 98 | |||
More Information
Set Membership: |
|

AGMO
Genomic Map