Assay Details
| Gene Symbol: TAFA3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015148485.2 | XP_015003971.1 | 668 | 95 | ||
| XM_028830327.1 | XP_028686160.1 | 1348 | 95 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

TAFA3| Gene Symbol: TAFA3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015148485.2 | XP_015003971.1 | 668 | 95 | ||
| XM_028830327.1 | XP_028686160.1 | 1348 | 95 | |||
Set Membership: |
|