Assay Details
| Gene Symbol: TAFA4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_028844354.1 | XP_028700187.1 | 300 | 130 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_028844354.1 transcripts.
Set Membership: |
|

TAFA4
Genomic Map