Assay Details
| Gene Symbol: TAFA4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015130436.2 | XP_014985922.1 | 944 | 146 | ||
| XM_028844354.1 | XP_028700187.1 | 557 | 146 | |||
| XM_028844355.1 | XP_028700188.1 | 639 | 146 | |||
| XM_028844356.1 | XP_028700189.1 | 421 | 146 | |||
| XM_028844357.1 | XP_028700190.1 | 322 | 146 | |||
More Information
Set Membership: |
|

TAFA4
Genomic Map