Assay Details
| Gene Symbol: PCGF6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001194611.2 | NP_001181540.1 | 1071 | 77 | ||
| XM_001113708.4 | XP_001113708.1 | 884 | 77 | |||
| XM_015148018.2 | XP_015003504.1 | 1105 | 77 | |||
| GenBank mRNA | JU494684.1 | - | 859 | 77 | ||
| JV724880.1 | - | 712 | 77 | |||
More Information
Set Membership: |
|

PCGF6
Genomic Map