Assay Details
| Gene Symbol: PRAMEF12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015109751.2 | XP_014965237.2 | 877 | 135 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

PRAMEF12| Gene Symbol: PRAMEF12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015109751.2 | XP_014965237.2 | 877 | 135 | ||
Set Membership: |
|