Assay Details
| Gene Symbol: P2RY10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001258158.1 | NP_001245087.1 | 137 | 143 | ||
| GenBank mRNA | JU473504.1 | - | 137 | 143 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001258158.1 transcripts.
Set Membership: |
|

P2RY10
Genomic Map