Assay Details
Gene Symbol: AMER2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001091613.4 | XP_001091613.1 | 1409 | 130 | ||
GenBank mRNA | JV574384.1 | - | 122 | 130 | ||
JV709405.1 | - | 367 | 130 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
