Assay Details
| Gene Symbol: CORO1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015113579.2 | XP_014969065.1 | 527 | 127 | ||
| XM_015113580.2 | XP_014969066.1 | 504 | 127 | |||
| XM_015113581.2 | XP_014969067.1 | 462 | 127 | |||
| GenBank mRNA | JU322535.1 | - | 282 | 127 | ||
| JU471529.1 | - | 308 | 127 | |||
| JV045804.1 | - | 285 | 127 | |||
| JV602029.1 | - | 308 | 127 | |||
More Information
Set Membership: |
|

CORO1B
Genomic Map