Assay Details
| Gene Symbol: FBLIM1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001084013.4 | XP_001084013.2 | 817 | 131 | ||
| XM_015119650.2 | XP_014975136.2 | 1047 | 131 | |||
| XM_028839315.1 | XP_028695148.1 | 1490 | 131 | |||
| XM_028839320.1 | XP_028695153.1 | 756 | 131 | |||
| GenBank mRNA | JV047450.1 | - | 720 | 131 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

FBLIM1
Genomic Map