Assay Details
| Gene Symbol: LOC702851 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015131474.2 | XP_014986960.1 | 2367 | 144 | ||
| XM_015131476.2 | XP_014986962.1 | 2228 | 144 | |||
| XM_015131477.2 | XP_014986963.1 | 2140 | 144 | |||
| GenBank mRNA | JU321664.1 | - | 1383 | 144 | ||
| JU327275.1 | - | 1346 | 144 | |||
| JV566823.1 | - | 1435 | 144 | |||
| JV667512.1 | - | 1428 | 144 | |||
| JV667513.1 | - | 1515 | 144 | |||
More Information
Set Membership: |
|

LOC702851
Genomic Map