Assay Details
| Gene Symbol: HVCN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001108107.4 | XP_001108107.1 | 891 | 60 | ||
| XM_015152790.2 | XP_015008276.1 | 459 | 60 | |||
| XM_028829866.1 | XP_028685699.1 | 978 | 60 | |||
| XM_028829867.1 | XP_028685700.1 | 757 | 60 | |||
| XM_028829868.1 | XP_028685701.1 | 766 | 60 | |||
| XM_028829869.1 | XP_028685702.1 | 668 | 60 | |||
| XM_028829870.1 | XP_028685703.1 | 1028 | 60 | |||
| XM_028829871.1 | XP_028685704.1 | 796 | 60 | |||
| GenBank mRNA | JU323431.1 | - | 320 | 60 | ||
| JU494828.1 | - | 284 | 60 | |||
More Information
Set Membership: |
|

HVCN1
Genomic Map