Assay Details
| Gene Symbol: PPAN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001204429.2 | NP_001191358.1 | 520 | 103 | ||
| XM_028838315.1 | XP_028694148.1 | 449 | 103 | |||
| GenBank mRNA | JU476436.1 | - | 397 | 103 | ||
| JU476437.1 | - | 397 | 103 | |||
| JV047678.1 | - | 407 | 103 | |||
| JV047679.1 | - | 407 | 103 | |||
| JV636276.1 | - | 384 | 103 | |||
More Information
Set Membership: |
|

PPAN
Genomic Map