Assay Details
| Gene Symbol: HEY1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001090360.4 | XP_001090360.2 | 377 | 84 | ||
| XM_015145608.2 | XP_015001094.1 | 377 | 84 | |||
| GenBank mRNA | JU322035.1 | - | 346 | 84 | ||
More Information
Set Membership: |
|

HEY1
Genomic Map