Assay Details
| Gene Symbol: CBR3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001193746.3 | NP_001180675.2 | 386 | 146 | ||
| GenBank mRNA | JU323428.1 | - | 412 | 146 | ||
| JV597154.1 | - | 231 | 146 | |||
| JV664450.1 | - | 244 | 146 | |||
More Information
Set Membership: |
|

CBR3
Genomic Map