Assay Details
| Gene Symbol: CBR3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001193746.3 | NP_001180675.2 | 492 | 104 | ||
| GenBank mRNA | JV597154.1 | - | 337 | 104 | ||
| JV664450.1 | - | 350 | 104 | |||
More Information
Set Membership: |
|

CBR3
Genomic Map