Assay Details
| Gene Symbol: COX10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001194644.2 | NP_001181573.1 | 728 | 120 | ||
| XM_015118624.2 | XP_014974110.2 | 731 | 120 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

COX10| Gene Symbol: COX10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001194644.2 | NP_001181573.1 | 728 | 120 | ||
| XM_015118624.2 | XP_014974110.2 | 731 | 120 | |||
Set Membership: |
|