Assay Details
| Gene Symbol: DHCR7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015113403.2 | XP_014968889.2 | 618 | 65 | ||
| XM_015113404.2 | XP_014968890.2 | 543 | 65 | |||
| XM_015113406.2 | XP_014968892.2 | 595 | 65 | |||
| XM_028833087.1 | XP_028688920.1 | 621 | 65 | |||
| GenBank mRNA | JU322693.1 | - | 521 | 65 | ||
| JV046340.1 | - | 464 | 65 | |||
| JV638000.1 | - | 523 | 65 | |||
More Information
Set Membership: |
|

DHCR7
Genomic Map