Assay Details
| Gene Symbol: FABP7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001257714.1 | NP_001244643.1 | 279 | 97 | ||
| XM_015137307.2 | XP_014992793.1 | 311 | 97 | |||
| GenBank mRNA | JU322666.1 | - | 279 | 97 | ||
More Information
Set Membership: |
|

FABP7
Genomic Map