Assay Details
| Gene Symbol: GDF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001115596.4 | XP_001115596.2 | 482 | 106 | ||
| GenBank mRNA | JV613055.1 | - | 433 | 106 | ||
| Gene Symbol: CERS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015123359.2 | XP_014978845.1 | 484 | 106 | ||
| XM_028838994.1 | XP_028694827.1 | 328 | 106 | |||
| GenBank mRNA | JV046740.1 | - | 426 | 106 | ||
| JV565100.1 | - | 427 | 106 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_001115596.4 transcripts.
Set Membership: |
|

GDF1
Genomic Map