Assay Details
| Gene Symbol: CERS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | JV046740.1 | - | 1660 | 88 | ||
| JV565100.1 | - | 1661 | 88 | |||
| Gene Symbol: GDF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001115596.4 | XP_001115596.2 | 1716 | 88 | ||
| GenBank mRNA | JV613055.1 | - | 1667 | 88 | ||
More Information
Set Membership: |
|

CERS1
Genomic Map