Assay Details
| Gene Symbol: APOBEC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_028829235.1 | XP_028685068.1 | 391 | 113 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

APOBEC1| Gene Symbol: APOBEC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_028829235.1 | XP_028685068.1 | 391 | 113 | ||
Set Membership: |
|